Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533782
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533782 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533782 Supplier Thermo Scientific Supplier No. PA533782
Add to Cart
Add to Cart

GPR88 Polyclonal Antibody

GPR88 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR88
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR88
Gene Accession No. Q9EPB7, Q9ESP4, Q9GZN0
Gene Alias AW061286; COCPMR; G protein coupled receptor 88; G protein-coupled receptor 88; Gpr88; G-protein coupled receptor 88; Probable G-protein coupled receptor 88; Strg; Striatum-specific G-protein coupled receptor
Gene Symbols GPR88
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human GPR88.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100346034, 100623311, 54112, 64378, 64443
Target Species Human, Mouse, Rat, Rabbit, Monkey, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.