Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR88 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1873 Supplier Bioss Supplier No. BS12017R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR88 is an Orphan-A GPCR with an unknown ligand. GPR88 expression has been documented almost exclusively in the striatum of the brain and not in any peripheral tissues. ESTs have been isolated from diseased brain, ear, and kidney libraries.

Specifications

Antigen GPR88
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR88
Gene Accession No. Q9GZN0
Gene Alias AW061286; BOS_3214; COCPMR; G protein coupled receptor 88; G protein-coupled receptor 88; GPR88; G-protein coupled receptor 88; G-protein coupled receptor 88-like; LOC100346034; Probable G-protein coupled receptor 88; STRG; Striatum-specific G-protein coupled receptor
Gene Symbols GPR88
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G protein coupled receptor 88.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54112
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.