Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR89A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533786
Description
GPR89A Polyclonal Antibody for IHC (P)
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifications
| GPR89A | |
| Polyclonal | |
| Unconjugated | |
| GPR89A | |
| 4933412D19Rik; CGI-13; G protein-coupled receptor 89; G protein-coupled receptor 89A; Golgi pH regulator; Golgi pH regulator A; Golgi pH regulator B; Gphr; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; Protein GPR89; Protein GPR89A; Protein GPR89B; Putative MAPK-activating protein PM01; putative NF-kappa-B-activating protein 90; SH120; STRG; UNQ192; UNQ192/PRO218 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100065612, 483157, 653519, 67549, 767859 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| B7ZAQ6, Q5BIM9, Q8BS95 | |
| Gpr89, GPR89A | |
| Synthetic 16 amino acid peptide from 3rd extracellular domain of human GPR89A. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Frog, Zebrafish, Bat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction