Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR89A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567875 Supplier Invitrogen™ Supplier No. PA567875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR89A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Water with 50% glycerol and 0.01% thimerosal; pH 7.4
Gene GPR89A
Gene Accession No. B7ZAQ6, Q8BS95
Gene Alias 4933412D19Rik; CGI-13; G protein-coupled receptor 89; G protein-coupled receptor 89A; Golgi pH regulator; Golgi pH regulator A; Golgi pH regulator B; Gphr; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; Protein GPR89; Protein GPR89A; Protein GPR89B; Putative MAPK-activating protein PM01; putative NF-kappa-B-activating protein 90; SH120; STRG; UNQ192; UNQ192/PRO218
Gene Symbols Gpr89, GPR89A
Host Species Rabbit
Immunogen Synthesized peptide derived from human GPR89A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653519, 67549
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.