Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR89B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA132840
Description
PA1-32840 detects GPCR SH120 from human samples.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifications
GPR89B | |
Polyclonal | |
Unconjugated | |
GPR89B | |
CGI-13; G protein-coupled receptor 89B; G protein-coupled receptor 89C; Golgi pH regulator A; Golgi pH regulator B; GPHR; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; protein GPR89; Protein GPR89A; Protein GPR89B; putative Golgi pH regulator C; putative MAPK-activating protein PM01; Putative NF-kappa-B-activating protein 90; putative protein GPR89C; SH120; UNQ192; UNQ192/PRO218 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51463 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P0CG08 | |
GPR89B | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human GPR89A. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction