Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR97 Antibody (2435C), PE/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10097PECY7
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Flow CytometrySpecifications
GPR97 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2435C | |
Flow Cytometry | |
G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession # Q86Y34 | |
0.1 mL | |
GPCR | |
222487 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction