Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR97 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116252
Description
GPR97 Polyclonal Antibody for Western Blot
GPR97 is expressed exclusively in pre-B and pre-T cells in the thymus but not in mature lymphoid or non-lymphoid tissues. ESTs have been isolated from spleen libraries. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Monkey (94%) Mouse, Rat (89%) Rabbit (83%)..
Specifications
GPR97 | |
Polyclonal | |
Unconjugated | |
ADGRG3 | |
ADGRG3; Adhesion G protein-coupled receptor G3; G protein-coupled receptor 97; GPR97; G-protein coupled receptor 97; G-protein coupled receptor PGR26; PB99; PGR26; probable G-protein coupled receptor 97 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
222487 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.50 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86Y34 | |
ADGRG3 | |
KLH conjugated synthetic peptide between 103-130 amino acids from the N-terminal region of human GPR97. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction