Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR98 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533792
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533792 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533792 Supplier Invitrogen™ Supplier No. PA533792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Gibbon, Monkey, Marmoset (95%); Elephant, Horse (89%); Dog, Panda (84%). Proteinase K treatment is required for antigen retrieval.

This gene encodes a member of the G-protein coupled receptor superfamily. The encoded protein contains a 7-transmembrane receptor domain, binds calcium and is expressed in the central nervous system. Mutations in this gene are associated with Usher syndrome 2 and familial febrile seizures. Several alternatively spliced transcripts have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR98
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRV1
Gene Accession No. Q8WXG9
Gene Alias 4-Feb; ADGRV1; ADGRV1 subunit alpha; ADGRV1 subunit beta; adhesion G protein-coupled receptor V1; Adhesion G-protein coupled receptor V1; FEB4; Frings; G protein-coupled receptor 98; Gpr98; G-protein coupled receptor 98; KIAA0686; KIAA1943; MASS1; Mgr1; monogenic; monogenic audiogenic seizure susceptibility protein 1; monogenic audiogenic seizure susceptibility protein 1 homolog; monogenic, audiogenic seizure susceptibility 1; monogenic, audiogenic seizure susceptibility 1 homolog; neurepin; USH2B; USH2C; Usher syndrome type-2C protein; very large G protein-coupled receptor 1; very large G-protein coupled receptor 1; VLGR1; VLGR1b
Gene Symbols ADGRV1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GPR98.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.