Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572830 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572830 Supplier Invitrogen™ Supplier No. PA572830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPRASP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRASP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GPRASP1
Gene Accession No. Q5JY77, Q5U4C1
Gene Alias 2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; KIAA0443; Per1 interacting protein; per1-interacting protein; Pips
Gene Symbols GPRASP1
Host Species Rabbit
Immunogen GPRASP1 antibody for a 15 amino acid peptide near the amino terminus of human GPRASP1. The immunogen is within amino acids 260 - 310 of GPRASP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67298, 9737
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.