Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPRASP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525727
Description
GPRASP1 Polyclonal Antibody for Western Blot, IHC (P), Flow
Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
Specifications
GPRASP1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5JY77 | |
GPRASP1 | |
KLH conjugated synthetic peptide between 1149-1176 amino acids from the C-terminal region of human GPRASP1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GPRASP1 | |
2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; KIAA0443; Per1 interacting protein; per1-interacting protein; Pips | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
9737 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction