Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRASP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553607 Supplier Invitrogen™ Supplier No. PA553607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPLVVRPKVR TQATTGARPK TETKSVPAAR PKTEAQAMSG ARPKTEVQVM GGARPKTEAQ GITGARPKTD ARAVGGARSK TDAKAIPGAR PKDEAQAWAQ SEFGTEAVSQ AEGVSQTNA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 72%.

The protein encoded by this gene is a member of a family that regulates the activity of G protein-coupled receptors (GPCRs). The encoded protein has been shown to be capable of interacting with several GPCRs, including the M1 muscarinic acetylcholine receptor and the calcitonin receptor. Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRASP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPRASP2
Gene Accession No. Q96D09
Gene Alias 5330440H13Rik; FLJ35662; FLJ37327; G protein-coupled receptor associated sorting protein 2; GASP2; GASP-2; GPRASP2; G-protein coupled receptor-associated sorting protein 2; prion protein ligand 5; Prpl5; RGD1561019
Gene Symbols GPRASP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPRASP2. Recombinant protein control fragment (Product #RP-91904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.