Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPRC5B Antibody (575926) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10253W
Description
GPRC5B Monoclonal antibody specifically detects GPRC5B in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
GPRC5B | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
575926 | |
Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein | |
NS0 mouse myeloma cell line transfected with human GPRC5B, Accession # Q9NZH0 | |
0.1 mL | |
GPCR | |
51704 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction