Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPRC5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533799
Description
GPRC5B Polyclonal Antibody for IHC (P)
This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
Specifications
| GPRC5B | |
| Polyclonal | |
| Unconjugated | |
| GPRC5B | |
| A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100052112, 100353261, 100856475, 293546, 516334, 51704, 64297 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q923Z0, Q9NZH0 | |
| GPRC5B | |
| Synthetic 18 amino acid peptide from N-terminal extracellular domain of human GPRC5B. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction