Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRC5C Polyclonal Antibody

Catalog No. PIPA132549
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132549 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132549 Supplier Invitrogen™ Supplier No. PA132549

Rabbit Polyclonal Antibody

PA1-32549 detects GPCR GPRC5C from human samples.

This polyclonal antibody was raised in Rabbit against GPRC5C. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPRC5C. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPRC5C
Gene Accession No. Q9NQ84
Gene Alias 1110028I06Rik; 3200002M13Rik; G protein-coupled receptor class C group 5 member C; G protein-coupled receptor, class C, group 5, member C; G protein-coupled receptor, family C, group 5, member C; Gprc5c; G-protein coupled receptor family C group 5 member C; MGC131820; orphan G-protein coupled receptor; PSEC0087; RAIG3; RAIG-3; retinoic acid inducible protein 3; retinoic acid responsive gene protein; retinoic acid-induced gene 3 protein
Gene Symbols GPRC5C
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human GPRC5C.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100060724, 483288, 55890
Target Species Human, Canine, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.