Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPRC5D Antibody (3D4), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | GPRC5D |
---|---|
Clone | 3D4 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for ELISASpecifications
GPRC5D | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
Recombinant Human GPRC5D protein [UniProt Q9NZD1] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3D4 | |
ELISA | |
Unconjugated | |
Human | |
GPCR | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
55507 | |
IgG4 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title