Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5D Mouse anti-Human, Unconjugated, Clone: 6D9, Abnova™

Catalog No. 89335121 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-121 Supplier Abnova Corporation Supplier No. H00055507M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for WB-Ti, S-ELISA, ELISA, WB-Re.

Specifications

Antigen GPRC5D
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6D9
Conjugate Unconjugated
Description G protein-coupled receptor, family C, group 5, member D
Gene GPRC5D
Gene Accession No. NM_018654
Gene Alias MGC129713, MGC129714
Host Species Mouse
Immunogen GPRC5D (NP_061124, 261 a.a. ∼ 345 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 55507
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.