Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPRC5D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570785
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined.
Specifications
GPRC5D | |
Polyclonal | |
Unconjugated | |
GPRC5D | |
G protein-coupled receptor class C group 5 member D; G protein-coupled receptor, class C, group 5, member D; G protein-coupled receptor, family C, group 5, member D; Gprc5d; G-protein coupled receptor family C group 5 member D; MGC129713; MGC129714; orphan G-protein coupled receptor; RGD1563887 | |
Rabbit | |
Affinity chromatography | |
RUO | |
93746 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9JIL6 | |
GPRC5D | |
synthetic peptide directed towards the C-terminal region of mouse GPRC5D. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction