Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRC5D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109785 Supplier Invitrogen™ Supplier No. PA5109785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILYRSCRQEC PLQGNACPVT AYQHSFQVEN QELSRARDSD GAEEDVALTS YGTPIQPQTV DPTQECFIPQ AKLSPQQDAG GV.

The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GPRC5D
Gene Accession No. Q9NZD1
Gene Alias G protein-coupled receptor class C group 5 member D; G protein-coupled receptor, class C, group 5, member D; G protein-coupled receptor, family C, group 5, member D; Gprc5d; G-protein coupled receptor family C group 5 member D; MGC129713; MGC129714; orphan G-protein coupled receptor; RGD1563887
Gene Symbols GPRC5D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 264-345 of human GPRC5D (NP_061124.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500349, 55507
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.