Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRC5D Recombinant Human Monoclonal Antibody (3D4)
GREENER_CHOICE

Catalog No. FEMA549973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549973 Supplier Invitrogen™ Supplier No. MA549973
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

GPRC5D Recombinant Monoclonal Antibody for ELISA

The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5D
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone 3D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene GPRC5D
Gene Accession No. Q9NZD1
Gene Alias G protein-coupled receptor class C group 5 member D; G protein-coupled receptor, class C, group 5, member D; G protein-coupled receptor, family C, group 5, member D; Gprc5d; G-protein coupled receptor family C group 5 member D; MGC129713; MGC129714; orphan G-protein coupled receptor; RGD1563887
Gene Symbols GPRC5D
Host Species Human
Immunogen Recombinant Human GPRC5D protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55507
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.