Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPRC6A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GPRC6A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GPRC6A Polyclonal specifically detects GPRC6A in Human, Mouse samples. It is validated for Western Blot.Specifications
GPRC6A | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
bA86F4.3, G protein-coupled receptor, family C, group 6, member A, GPCR, G-protein coupled receptor family C group 6 member A, G-protein coupled receptor GPCR33, hGPCR33, hGPRC6A, predicted with SOSUI analysis, seven transmembrane helix receptor | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-590 of human GPRC6A (NP_683766.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS with 50% glycerol, pH7.3. | |
222545 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title