Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GPRIN1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1446MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1446MI 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1446MI Supplier MilliporeSigma™ Supplier No. ABN1446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G protein-regulated inducer of neurite outgrowth 1 (UniProt Q7Z2K8; also known as GRIN1) is encoded by the GPRIN1 (also known as KIAA1893) gene (Gene ID 114787) in human. Originally identified as a binding protein for guanosine 5&prime-3-O-(thio)triphosphate-bound G(z)alphaz, G protein-regulated inducer of neurite outgrowth 1 (GPRIN1) is a signaling molecule that functions as a downstream target of G(o)alpha. When co-expressed with constitutively active form of G(o)alpha, GPRIN1 is demonstrated to stimulate neurite extensions in Neuro2a cells. Gprin1 expression in mice is limited to the nervous system at all developmental stages, occurring within the entire embryonic mantle zones and becoming restricted to sets of nuclei at postnatal to adult stages. Gprin1 immunoreactivity is found colocalized with G(o)alpha at neuronal dendrites and axons, while being undetected in glial cells. Studies conducted in rat reveals that 7 nicotinic receptor ( 7) significantly reduces axon growth in hippocampal neurons by coupling to Gprin1, which targets it to the growth cone. RNAi-mediated Gprin1 knockdown abolishes the localization and calcium signaling of 7 at the growth cone. The 7/Gprin1 interaction appears intimately linked to a G(o)alpha, growth-associated protein 43, and CDC42 cytoskeletal regulatory pathway within the developing axon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRIN1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene GPRIN1, KIAA1893
Gene Accession No. Q7Z2K8
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an internal sequence of human GPRIN1.
Purification Method Affinity Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_443131
Test Specificity This rabbit polyclonal antibody targets an internal sequence present in both spliced isoforms of human GPRIN1 protein reported by UniProt (Q7Z2K8).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.