Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104272 Supplier Invitrogen™ Supplier No. PA5104272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GRIN2.

May be involved in neurite outgrowth. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRIN2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPRIN2
Gene Accession No. O60269
Gene Alias C130040D06Rik; C230073P13; G protein regulated inducer of neurite outgrowth 2; G protein-regulated inducer of neurite outgrowth 2; Gm286; GPRIN2; GRIN2; KIAA0514; mKIAA0514; RGD1308922
Gene Symbols GPRIN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPRIN2(Accession O60269), corresponding to amino acid residues A139-L189.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9721
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.