Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPS1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518160
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
Specifications
| GPS1 | |
| Polyclonal | |
| Unconjugated | |
| GPS1 | |
| COP9 signalosome complex subunit 1; Cops1; CSN1; G protein pathway suppressor 1; Gps1; GPS-1; JAB1-containing signalosome subunit 1; MFH; MFH protein; MGC71287; Protein MFH; R75577; SGN1; signalosome subunit 1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 2873 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13098 | |
| GPS1 | |
| Synthetic peptide sequence (PANSQSRMSTNM) corresponding to the C-terminus amino acids of GPS1 (aa 480-491). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction