Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527149
Description
Recommended positive controls: HeLaS3, Molt-4, 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Chicken (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
Specifications
GPS1 | |
Polyclonal | |
Unconjugated | |
GPS1 | |
COP9 signalosome complex subunit 1; Cops1; CSN1; G protein pathway suppressor 1; Gps1; GPS-1; JAB1-containing signalosome subunit 1; MFH; MFH protein; MGC71287; Protein MFH; R75577; SGN1; signalosome subunit 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117039, 209318, 2873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P97834, Q13098, Q99LD4 | |
GPS1 | |
Synthetic peptide corresponding to a region within amino acids 430 and 491 of Human CSN1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction