Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578650 Supplier Invitrogen™ Supplier No. PA578650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Human (100%), Rat (97%), Zebrafish (91%), Xenopus laevis (93%), Chicken (95%), Chimpanzee (98%), Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene GPS1
Gene Accession No. Q13098, Q99LD4
Gene Alias COP9 signalosome complex subunit 1; Cops1; CSN1; G protein pathway suppressor 1; Gps1; GPS-1; JAB1-containing signalosome subunit 1; MFH; MFH protein; MGC71287; Protein MFH; R75577; SGN1; signalosome subunit 1
Gene Symbols GPS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CSN1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209318, 2873
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.