Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566932 Supplier Invitrogen™ Supplier No. PA566932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHGHFQPTQTG FLQPGGALSL QKQMEHANQQ TGFSDSSSLR PMHPQALHPA PGLLASPQLP VQMQPAGKSG FAATSQPGPR LPFIQHSQNP RFYHK Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

This gene encodes a protein involved in G protein-mitogen-activated protein kinase (MAPK) signaling cascades. When overexpressed in mammalian cells, this gene could potently suppress a RAS- and MAPK-mediated signal and interfere with JNK activity, suggesting that the function of this gene may be signal repression. The encoded protein is an integral subunit of the NCOR1-HDAC3 (nuclear receptor corepressor 1-histone deacetylase 3) complex, and it was shown that the complex inhibits JNK activation through this subunit and thus could potentially provide an alternative mechanism for hormone-mediated antagonism of AP1 (activator protein 1) function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPS2
Gene Accession No. Q13227
Gene Alias AI505953; AMF-1; G protein pathway suppressor 2; GPS2; GPS-2
Gene Symbols GPS2
Host Species Rabbit
Immunogen Recombinant Human CSN2. Recombinant protein control fragment (Product #RP-107585).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.