Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPSM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568196
Description
Predicted to react with Mouse and Rat samples.
G-protein signaling modulators (GPSMs) play diverse functional roles through their interaction with G-protein subunits. This gene encodes a receptor-independent activator of G protein signaling, which is one of several factors that influence the basal activity of G-protein signaling systems. The protein contains seven tetratricopeptide repeats in its N-terminal half and four G-protein regulatory (GPR) motifs in its C-terminal half. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
GPSM1 | |
Polyclonal | |
Unconjugated | |
GPSM1 | |
1810037C22Rik; Activator of G-protein signaling 3; Ags3; AW107933; C10a; DKFZP727I051; G-protein signaling modulator 1; G-protein signaling modulator 1 (AGS3-like, C. elegans); G-protein signalling modulator 1 (AGS3-like, C. elegans); G-protein-signaling modulator 1; Gpsm1 | |
Rabbit | |
affinity chromatography | |
RUO | |
246254, 26086, 67839 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6IR34, Q86YR5, Q9R080 | |
GPSM1 | |
Synthesized peptide derived from human GPSM1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction