Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPSM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559911 Supplier Invitrogen™ Supplier No. PA559911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPAASEKPDL AGYEAQGARP KRTQRLSAET WDLLRLPLER EQNGDSHHSG DWRGPSRDSL PLPVRSRK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

G-protein signaling modulators (GPSMs) play diverse functional roles through their interaction with G-protein subunits. This gene encodes a receptor-independent activator of G protein signaling, which is one of several factors that influence the basal activity of G-protein signaling systems. The protein contains seven tetratricopeptide repeats in its N-terminal half and four G-protein regulatory (GPR) motifs in its C-terminal half. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPSM1
Gene Accession No. Q86YR5
Gene Alias 1810037C22Rik; Activator of G-protein signaling 3; Ags3; AW107933; C10a; DKFZP727I051; G-protein signaling modulator 1; G-protein signaling modulator 1 (AGS3-like, C. elegans); G-protein signalling modulator 1 (AGS3-like, C. elegans); G-protein-signaling modulator 1; Gpsm1
Gene Symbols GPSM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPSM1. Recombinant protein control fragment (Product #RP-98515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.