Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPSM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518646
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518646 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518646 Supplier Invitrogen™ Supplier No. PA518646
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The protein encoded by this gene belongs to a family of proteins that modulate activation of G proteins, which transduce extracellular signals received by cell surface receptors into integrated cellular responses. The N-terminal half of this protein contains 10 copies of leu-gly-asn repeat, and the C-terminal half contains 4 GoLoco motifs, which are involved in guanine nucleotide exchange. This protein may play a role in neuroblast division and in the development of normal hearing. Mutations in this gene are associated with autosomal recessive nonsyndromic deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSM2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GPSM2
Gene Accession No. P81274
Gene Alias 6230410J09Rik; CMCS; DFNB82; G protein signaling modulator 2; G-protein signaling modulator 2; G-protein signaling modulator 2 (AGS3-like, C. elegans); G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; GPSM2; HGNC:29501; hypothetical protein MGC63574; LGN; LGN protein; Mosaic protein LGN; Pins; Pins homolog; pins homolog (Drosophia); Pinsa; RGD1560967; TPR-motif and GoLoco-motif containing protein; zgc:63574
Gene Symbols GPSM2
Host Species Goat
Immunogen Synthetic peptide sequence (NRLKGKKYKTNSSTK) corresponding to the internal amino acids of GPSM2 (aa 448-463).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29899
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.