Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPSM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587981 Supplier Thermo Scientific Supplier No. PA587981
Add to Cart
Add to Cart

GPSM2 Polyclonal Antibody

GPSM2 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GPSM2
Gene Accession No. P81274, Q8VDU0
Gene Alias 6230410J09Rik; CMCS; DFNB82; G protein signaling modulator 2; G-protein signaling modulator 2; G-protein signaling modulator 2 (AGS3-like, C. elegans); G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; GPSM2; HGNC:29501; hypothetical protein MGC63574; LGN; LGN protein; Mosaic protein LGN; Pins; Pins homolog; pins homolog (Drosophia); Pinsa; RGD1560967; TPR-motif and GoLoco-motif containing protein; zgc:63574
Gene Symbols GPSM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 380-490 of human GPSM2 (NP_0374283).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29899, 76123
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.