Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPSM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568197 Supplier Invitrogen™ Supplier No. PA568197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Interacts with subunit of G(i) alpha proteins and regulates the activation of G(i) alpha proteins. Target protein is a member of a family of proteins able to activate G-protein signalling in the absence of a receptor; human homolog is located within the MHC class III region; may play a role in immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSM3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPSM3
Gene Accession No. Q3U1Z5, Q6MG88, Q9Y4H4
Gene Alias AA960287; activator of G-protein signaling 4; activator of G-protein signalling 4; AGS4; C6orf9; G18; G18.1a; G18.1b; G18.2; G-protein signaling modulator 3; G-protein signaling modulator 3 (AGS3-like, C. elegans); G-protein signalling modulator 3 (AGS3-like, C. elegans); G-protein-signaling modulator 3; GPSM3; Ng1; Protein G18
Gene Symbols GPSM3
Host Species Rabbit
Immunogen Synthesized peptide derived from human GPSM3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106512, 406163, 63940
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.