Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPSN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543178 Supplier Invitrogen™ Supplier No. PA543178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PFIYGHKYDF TSSRHTVVHL ACICHSFHYI KRLLETLFVH RFSHGTMPLR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TECR
Gene Accession No. Q9NZ01
Gene Alias 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; glycoprotein, synaptic 2; GPSN2; hypothetical protein LOC614105; MRT14; neuroprotective protein 13; SC2; Synaptic glycoprotein SC2; Tecr; TER; trans-2,3-enoyl-CoA reductase; trans-2,3-enoyl-CoA reductase; very-long-chain enoyl-CoA reductase; very-long-chain enoyl-CoA reductase
Gene Symbols TECR
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GPSN2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9524
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.