Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPSN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563311 Supplier Invitrogen™ Supplier No. PA563311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKHYEVEILD AKTREKLCFL DKVEPHATIA EIKNLF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TECR
Gene Accession No. Q9NZ01
Gene Alias 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; glycoprotein, synaptic 2; GPSN2; hypothetical protein LOC614105; MRT14; neuroprotective protein 13; SC2; Synaptic glycoprotein SC2; Tecr; TER; trans-2,3-enoyl-CoA reductase; trans-2,3-enoyl-CoA reductase; very-long-chain enoyl-CoA reductase; very-long-chain enoyl-CoA reductase
Gene Symbols TECR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPSN2. Recombinant protein control fragment (Product #RP-104286).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9524
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.