Learn More
Invitrogen™ GPSN2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total GPSN2.
Specifications
Specifications
| Antigen | GPSN2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TECR |
| Gene Accession No. | Q64232, Q9CY27, Q9NZ01 |
| Gene Alias | 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; glycoprotein, synaptic 2; GPSN2; hypothetical protein LOC614105; MRT14; neuroprotective protein 13; SC2; Synaptic glycoprotein SC2; Tecr; TER; trans-2,3-enoyl-CoA reductase; trans-2,3-enoyl-CoA reductase; very-long-chain enoyl-CoA reductase; very-long-chain enoyl-CoA reductase |
| Gene Symbols | TECR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.