Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPT Monoclonal Antibody (C4), Invitrogen™

Catalog No. FEMA550914
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550914 20 μL
FEMA550913 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550914 Supplier Invitrogen™ Supplier No. MA550914
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene GPT
Gene Accession No. P24298, P25409
Gene Alias 1300007J06Rik; 2310022B03Rik; AAT1; alanine aminotransferase; alanine aminotransferase 1; ALT; ALT1; glutamate pyruvate transaminase 1; glutamic pyruvate transaminase; glutamic pyruvic transaminase (alanine aminotrasferase); glutamic pyruvic transaminase 1, soluble; glutamic pyruvic transaminase, soluble; glutamic-alanine transaminase 1; glutamic--alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); glutamic--pyruvic transaminase; glutamic--pyruvic transaminase 1; GPT; GPT 1; Gpt1; Gpt-1
Gene Symbols GPT
Host Species Mouse
Immunogen Recombinant Human ALT protein, Ile255-Thr492 + LEHHHHHH (Accession #P24298).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2875, 81670
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.