Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583178 Supplier Invitrogen™ Supplier No. PA583178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASSTGDRSQ AVRNGLRAKV LTLDGMNPRV RRVEYAVRGP IVQRALELEQ ELRQGVKKPF T Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 85%.

GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPT
Gene Accession No. P24298, P25409, Q8QZR5
Gene Alias 1300007J06Rik; 2310022B03Rik; AAT1; alanine aminotransferase; alanine aminotransferase 1; ALT; ALT1; glutamate pyruvate transaminase 1; glutamic pyruvate transaminase; glutamic pyruvic transaminase (alanine aminotrasferase); glutamic pyruvic transaminase 1, soluble; glutamic pyruvic transaminase, soluble; glutamic-alanine transaminase 1; glutamic--alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); glutamic--pyruvic transaminase; glutamic--pyruvic transaminase 1; GPT; GPT 1; Gpt1; Gpt-1
Gene Symbols GPT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPT. Recombinant protein control fragment (Product #RP-93886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2875, 76282, 81670
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.