Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543270 Supplier Invitrogen™ Supplier No. PA543270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PEYSSNVELA SFHSTSKGYM GECGYRGGYM EVINLHPEIK GQLVKLLSVR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

GPT and GPT2 (EC 2.6.1.2), also known as alanine transaminases, are pyridoxal enzymes that catalyze the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. By mediating the conversion of these 4 major intermediate metabolites, these transaminases have roles in gluconeogenesis and in amino acid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GPT2
Gene Accession No. Q8TD30
Gene Alias 4631422C05Rik; AAT2; Alanine aminotransferase 2; ALT2; AU014768; AU041193; C87201; Cc2-5; glutamate pyruvate transaminase 2; glutamic pyruvate transaminase (alanine aminotransferase) 2; glutamic pyruvate transaminase 2; glutamic--alanine transaminase 2; glutamic-pyruvate transaminase 2; Glutamic--pyruvic transaminase 2; GPT 2; GPT2; im:7150662; LOW QUALITY PROTEIN: alanine aminotransferase 2; MRT49; sb:cb580; zgc:165657
Gene Symbols GPT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GPT2 (aa 325-374).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84706
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.