Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPVI Antibody (784808), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6758PEATT594
Description
GPVI Monoclonal antibody specifically detects GPVI in Mouse samples. It is validated for Flow CytometrySpecifications
GPVI | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51206 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
784808 | |
Flow Cytometry | |
Glycoprotein 6, glycoprotein VI (platelet), GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI | |
Mouse myeloma cell line NS0-derived recombinant mouse GPVI, Gly24-Lys265, Accession # P0C191 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction