Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX2 Recombinant Rabbit Monoclonal Antibody (HL125)
GREENER_CHOICE

Catalog No. PIMA536260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536260 Supplier Invitrogen™ Supplier No. MA536260
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: HepG2.

This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL125
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GPX2
Gene Accession No. P18283, P83645, Q9JHC0
Gene Alias gastrointestinal glutathione peroxidase; GI-GPx; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP; GPRP-2; GPX2; GPx-2; GPX-GI; GSHPx-2; GSHPX-GI; HGNC:4554; intestinal GPx
Gene Symbols GPX2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPX2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14776, 2877, 29326
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.