Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145089 Supplier Invitrogen™ Supplier No. PA5145089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAFIAKSFYD LSAISLDGEK VDFNTFRGRA VLIENVAS Antigen sequence identity: Mouse 89%; Rat 92%.

This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPX2
Gene Accession No. P18283
Gene Alias gastrointestinal glutathione peroxidase; GI-GPx; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP; GPRP-2; GPX2; GPx-2; GPX-GI; GSHPx-2; GSHPX-GI; HGNC:4554; intestinal GPx
Gene Symbols GPX2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-38. Recombinant protein control fragment (Product #RP-110129).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2877
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.