Learn More
Invitrogen™ GPX3 Polyclonal Antibody

Description
The target sequence has 89% sequence homology with mouse, 88% sequence homology with bovine and 84% sequence homology with porcine. Suggested positive control: HeLa whole cell extract.
Specifications
Specifications
| Antigen | GPX3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.1% sodium azide |
| Gene | Gpx3 |
| Gene Accession No. | P22352, P23764 |
| Gene Alias | AA960521; EGPx; extracellular glutathione peroxidase; extracellular GPx; glutathione peroxidase 3; glutathione peroxidase 3 (plasma); GPx; Gpx3; GPx-3; Gpxp; GPx-P; GSHPx-3; GSHPx-P; plasma form; plasma glutathione peroxidase; plasma GPx |
| Gene Symbols | Gpx3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.