Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPX4 Mouse anti-Human, CoraLite 488, Clone: 3F5G5, Proteintech
SDP

Catalog No. 502257410 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7410 Supplier Proteintech Group Inc Supplier No. CL48867763100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPX4 (Phospholipid hydroperoxide glutathione peroxidase, mitochondrial) protects cells against membrane lipid peroxidation and cell death. Required for normal sperm development and male fertility.It has two isforms about 20KDa and 22KDa, respectively. GPX4 is a monomer,but it has a tendency to form higher mass oligomers (PMID:17630701). It presents primarily in testis.

Specifications

Antigen GPX4
Applications Immunofluorescence
Classification Monoclonal
Clone 3F5G5
Concentration 1000 μg/mL
Conjugate CoraLite 488
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2879
Target Species Human, Non-human Primate
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.