Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | GPX4 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119142
|
Invitrogen™
PA5119142 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121951
|
Invitrogen™
PA5121951 |
20 μL |
Each for $209.50
|
|
|||||
Description
GPX4 Polyclonal Antibody for Western Blot, IHC (P)
Glutathione peroxidase catalyzes the reduction of hydrogen peroxide, organic hydroperoxide, and lipid peroxides by reduced glutathione and functions in the protection of cells against oxidative damage. Human plasma glutathione peroxidase has been shown to be a selenium-containing enzyme and the UGA codon is translated into a selenocysteine. Through alternative splicing and transcription initiation, rat produces proteins that localize to the nucleus, mitochondrion, and cytoplasm. In humans, experimental evidence for alternative splicing exists; alternative transcription initiation and the cleavage sites of the mitochondrial and nuclear transit peptides need to be experimentally verified.Specifications
GPX4 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
GPX4 | |
glutathione peroxidase 4; GP-GPx4; Gpx4; gpx-4; GSHPx-4; HGNC:4556; MCSP; mtPHGPx; Phgpx; phospholipid hydroperoxidase; phospholipid hydroperoxide glutathione peroxidase; phospholipid hydroperoxide glutathione peroxidase, mitochondrial; phospholipid hydroperoxide glutathione peroxidase, nuclear; SMDS; snGPx; snPHGPx; sperm nuclei glutathione peroxidase; sperm nucleus glutathione peroxidase | |
GPX4 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P36969 | |
2879 | |
Recombinant protein Glutathione Peroxidase 4. The antigen corresponds to amino acid range 74-197 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title