Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX4 Recombinant Rabbit Monoclonal Antibody (9D4)
GREENER_CHOICE

Catalog No. FEMA549854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549854 Supplier Invitrogen™ Supplier No. MA549854
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GPX4 Recombinant Monoclonal Antibody for ELISA, IHC (P)

Glutathione peroxidase catalyzes the reduction of hydrogen peroxide, organic hydroperoxide, and lipid peroxides by reduced glutathione and functions in the protection of cells against oxidative damage. Human plasma glutathione peroxidase has been shown to be a selenium-containing enzyme and the UGA codon is translated into a selenocysteine. Through alternative splicing and transcription initiation, rat produces proteins that localize to the nucleus, mitochondrion, and cytoplasm. In humans, experimental evidence for alternative splicing exists; alternative transcription initiation and the cleavage sites of the mitochondrial and nuclear transit peptides need to be experimentally verified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX4
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 9D4
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPX4
Gene Accession No. P36969
Gene Alias glutathione peroxidase 4; GP-GPx4; Gpx4; gpx-4; GSHPx-4; HGNC:4556; MCSP; mtPHGPx; Phgpx; phospholipid hydroperoxidase; phospholipid hydroperoxide glutathione peroxidase; phospholipid hydroperoxide glutathione peroxidase, mitochondrial; phospholipid hydroperoxide glutathione peroxidase, nuclear; SMDS; snGPx; snPHGPx; sperm nuclei glutathione peroxidase; sperm nucleus glutathione peroxidase
Gene Symbols GPX4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPX4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2879
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.