Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102342 Supplier Invitrogen™ Supplier No. PA5102342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPX5.

Protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione. May constitute a glutathione peroxidase-like protective system against peroxide damage in sperm membrane lipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPX5
Gene Accession No. O75715, P21765, P30710
Gene Alias Arep; arMEP24; AV379049; EGLP; epididymal androgen-related protein; epididymal secretory glutathione peroxidase; epididymis secretory sperm binding protein Li 75p; epididymis-specific glutathione peroxidase-like protein; Glutathione peroxidase 5; Gpx5; GPx-5; GSHPx-5; HEL-S-75p; lysosomal trafficking regulator, long splice form; major androgen-regulated protein; OTTHUMP00000017919; OTTHUMP00000214636
Gene Symbols GPX5
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPX5(Accession O75715), corresponding to amino acid residues L82-F132.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113919, 14780, 2880
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.