Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX6 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $539.50

Specifications

Antigen GPX6
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119143
View Documents
Invitrogen™
PA5119143
200 μL
Each for $539.50
Only null left
Add to Cart
 
PIPA5121950
View Documents
Invitrogen™
PA5121950
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

GPX6 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene belongs to the glutathione peroxidase family, members of which catalyze the reduction of hydrogen peroxide, organic hydroperoxides and lipid hydroperoxides, and thereby protect cells against oxidative damage. Several isozymes of this gene family exist in vertebrates, which vary in cellular location and substrate specificity. Expression of this gene has been observed in embryos and olfactory epithelium; however, the exact function of this gene is not known. This isozyme is a selenoprotein in humans, containing the rare amino acid selenocysteine (Sec) at its active site. Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal. The orthologs of this gene in mouse and rat (and some other species) contain a cysteine (Cys) residue in place of the Sec residue, and their corresponding mRNAs lack SECIS element.
Specifications

Specifications

GPX6
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Rat
Gpx6
Glutathione Peroxidase 6; Glutathione Peroxidase 6 Olfactory
Gpx6
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q64625
259233
Recombinant protein Glutathione Peroxidase 6, Olfactory. The antigen corresponds to amino acid range 20-221 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.