Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPX7 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518546
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
Specifications
GPX7 | |
Polyclonal | |
Unconjugated | |
GPX7 | |
3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2882 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96SL4 | |
GPX7 | |
Synthetic peptide sequence (CNQFGQQEPDSNK) corresponding to the internal amino acids of GPX7 (aa 86-99). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction