Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528854 Supplier Invitrogen™ Supplier No. PA528854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Mouse embryonic fibroblast. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX7
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene GPX7
Gene Accession No. Q96SL4, Q99LJ6
Gene Alias 3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828
Gene Symbols GPX7
Host Species Rabbit
Immunogen Recombinant protein corresponding to human GPX7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2882, 67305
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.