Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120875 Supplier Invitrogen™ Supplier No. PA5120875
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, LO2, SW480, Mouse brain, Mouse heart. The target is usually found in the following locations: Secreted. Immunogen sequence: QQEQDFYDFK AVNIRGKLVS LEKYRGSVSL VVNVASECGF TDQHYRALQQ LQRDLGPHHF NVLAFPCNQF GQQEPDSNKE IESFARRTYS VSFPMFSKIA VTGTGAHPAF KYLAQTSGKE PTWNFWKYLV APDGKVVGAW DPTVSVEEVR PQITALVRKL ILLKREDL.

Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPX7
Gene Accession No. Q96SL4, Q99LJ6
Gene Alias 3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828
Gene Symbols GPX7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-187 of human GPX7 (NP_056511.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2882, 298376, 67305
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.