Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPX8 Antibody (8-1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313659
This item is not returnable.
View return policy
Description
GPX8 Monoclonal antibody specifically detects GPX8 in Human samples. It is validated for Western BlotSpecifications
| GPX8 | |
| Monoclonal | |
| Unconjugated | |
| EC 1.11.1, EC 1.11.1.9, EPLA847, glutathione peroxidase 8 (putative), GPx-8, GSHPx-8, probable glutathione peroxidase 8, UNQ847 | |
| The immunogen used to generate this antibody corresponds to human GPX8. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 2023-08-01 00:00:00 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 493869 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction